Skip to Content
Merck
All Photos(1)

Key Documents

KEY465196821

Key Organics / BIONET

butyl 6-chloro-2H-chromene-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
266.72
MDL number:
Pricing and availability is not currently available.

Iupac Name

butyl 6-chloro-2H-chromene-3-carboxylate

Inchi Code

1S/C14H15ClO3/c1-2-3-6-17-14(16)11-7-10-8-12(15)4-5-13(10)18-9-11/h4-5,7-8H,2-3,6,9H2,1H3

InChI key

ILRPQVSQESAMSQ-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/5G-301S

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/5G-301S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library