Skip to Content
Merck
All Photos(1)

Key Documents

KEY003446756

Key Organics / BIONET

4-chloro-1-methyl-[1,2,4]triazolo[4,3-a]quinoxaline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
218.65
MDL number:

mp

222 - 224

Iupac Name

4-chloro-1-methyl[1,2,4]triazolo[4,3-a]quinoxaline

Inchi Code

1S/C10H7ClN4/c1-6-13-14-10-9(11)12-7-4-2-3-5-8(7)15(6)10/h2-5H,1H3

InChI key

KJXIBZROUGENCF-UHFFFAOYSA-N

storage temp.

Store under inert gas at 4C

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/12E-939

Melting Point

222 - 224

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/12E-939


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library