Skip to Content
Merck
All Photos(1)

Key Documents

ENAH95E7403D

Enamine

rac-(2R,3R)-3-({[(tert-butoxy)carbonyl]amino}methyl)piperidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
258.32
MDL number:
Pricing and availability is not currently available.

Iupac Name

(2S,3S)-3-(((tert-butoxycarbonyl)amino)methyl)piperidine-2-carboxylic acid

Inchi Code

1S/C12H22N2O4/c1-12(2,3)18-11(17)14-7-8-5-4-6-13-9(8)10(15)16/h8-9,13H,4-7H2,1-3H3,(H,14,17)(H,15,16)/t8-,9-/m0/s1

InChI key

HYZWKQAIIIURNA-IUCAKERBSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-27100695

MSDS

https://www.enaminestore.com/catalog/EN300-27100695


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library