ENAH95E7388E
Enamine
rac-(1R,4S,5R,6R)-2-oxa-3-azabicyclo[2.2.2]octane-5,6-diol hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(1S,4R,5S,6S)-2-oxa-3-azabicyclo[2.2.2]octane-5,6-diol hydrochloride
Inchi Code
1S/C6H11NO3.ClH/c8-5-3-1-2-4(6(5)9)10-7-3;/h3-9H,1-2H2;1H/t3-,4+,5+,6-;/m1./s1
InChI key
KRHQIFIWPADYMB-OLALXQGDSA-N
storage temp.
4
Salt Data
Cl
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-7353104
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-7353104
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.