ENAH5802DF77
Enamine
methyl N-[4-(bromomethyl)phenyl]carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl (4-(bromomethyl)phenyl)carbamate
Inchi Code
1S/C9H10BrNO2/c1-13-9(12)11-8-4-2-7(6-10)3-5-8/h2-5H,6H2,1H3,(H,11,12)
InChI key
XUKIVPRPJHTDHK-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-7352627
MSDS
https://www.enaminestore.com/catalog/EN300-7352627
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.