ENAH304D7EEC
Enamine
2-amino-4-methylpentane-1-sulfonyl fluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
183.25
Iupac Name
2-amino-4-methylpentane-1-sulfonyl fluoride
Inchi Code
1S/C6H14FNO2S/c1-5(2)3-6(8)4-11(7,9)10/h5-6H,3-4,8H2,1-2H3
InChI key
VSOSLMOKKGXDCD-UHFFFAOYSA-N
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-1576971
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.