ENAH11E0D51B
Enamine
tert-butyl N-[1-(chloromethyl)cyclobutyl]carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl (1-(chloromethyl)cyclobutyl)carbamate
Inchi Code
1S/C10H18ClNO2/c1-9(2,3)14-8(13)12-10(7-11)5-4-6-10/h4-7H2,1-3H3,(H,12,13)
InChI key
XOWWOCWENCBRHV-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-1880582
MSDS
https://www.enaminestore.com/catalog/EN300-1880582
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.