Skip to Content
Merck
All Photos(1)

Key Documents

ENAH0424BACA

Enamine

ethyl 2-[(1E)-2,3-dihydro-1H-inden-1-ylidene]acetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
202.25
MDL number:

Iupac Name

ethyl (2E)-2,3-dihydro-1H-inden-1-ylideneethanoate

Inchi Code

1S/C13H14O2/c1-2-15-13(14)9-11-8-7-10-5-3-4-6-12(10)11/h3-6,9H,2,7-8H2,1H3/b11-9+

InChI key

KRGWQDMTJGWQFM-PKNBQFBNSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1460258

MSDS

https://www.enaminestore.com/catalog/EN300-1460258


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library