Skip to Content
Merck
All Photos(1)

Key Documents

ENA964473697

Enamine

ethyl 2,2-difluoro-2-(4-hydroxyoxan-4-yl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
224.2
MDL number:
Pricing and availability is not currently available.

Iupac Name

ethyl difluoro(4-hydroxytetrahydro-2H-pyran-4-yl)acetate

Inchi Code

1S/C9H14F2O4/c1-2-15-7(12)9(10,11)8(13)3-5-14-6-4-8/h13H,2-6H2,1H3

InChI key

HKBJQIJEGIPIHT-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-706064

MSDS

https://www.enaminestore.com/catalog/EN300-706064


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library