ENA428072745
Enamine
N2-benzyl-1,3,4-oxadiazole-2,5-diamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
115-117
Iupac Name
N~2~-benzyl-1,3,4-oxadiazole-2,5-diamine
Inchi Code
1S/C9H10N4O/c10-8-12-13-9(14-8)11-6-7-4-2-1-3-5-7/h1-5H,6H2,(H2,10,12)(H,11,13)
InChI key
GHUNXWZWFGGJJR-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-175566
Melting Point
115-117
MSDS
https://www.enaminestore.com/catalog/EN300-175566
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service