Skip to Content
Merck
All Photos(1)

Key Documents

COM448633921

Combi-Blocks, Inc.

2-Piperazinopyridine-3-boronic acid, pinacol ester dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
362.11
MDL number:
Pricing and availability is not currently available.

Iupac Name

1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]piperazine dihydrochloride

Inchi Code

1S/C15H24BN3O2.2ClH/c1-14(2)15(3,4)21-16(20-14)12-6-5-7-18-13(12)19-10-8-17-9-11-19;;/h5-7,17H,8-11H2,1-4H3;2*1H

InChI key

JRSUYHYCYIQOQE-UHFFFAOYSA-N

storage temp.

Freezer

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?PN-4513

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?PN-4513


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library