COM448622732
Combi-Blocks, Inc.
3-[4-Fluoro-3-(trifluoromethyl)phenyl]aniline hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4'-fluoro-3'-(trifluoromethyl)[1,1'-biphenyl]-3-amine hydrochloride
Inchi Code
1S/C13H9F4N.ClH/c14-12-5-4-9(7-11(12)13(15,16)17)8-2-1-3-10(18)6-8;/h1-7H,18H2;1H
InChI key
XKWXAYVIPJOYBB-UHFFFAOYSA-N
Purity
98%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0513
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0513
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.