Skip to Content
Merck
All Photos(1)

Key Documents

CH6371382917

Chem Impex International, Inc.

2-(3,4-Dihydro-2H-quinolin-1-yl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176.26
MDL number:

Iupac Name

2-(3,4-dihydro-1(2H)-quinolinyl)ethanamine

Inchi Code

1S/C11H16N2/c12-7-9-13-8-3-5-10-4-1-2-6-11(10)13/h1-2,4,6H,3,5,7-9,12H2

InChI key

ALWARSIRJIEZMK-UHFFFAOYSA-N

storage temp.

Store at 0-8C

Purity

95%

Shipping Temperature

RT

Physical Form

Light brown solid

Product Link

https://www.chemimpex.com/product/productinfo/2-3-4-dihydro-2-i-h-i-quinolin-1-yl-ethylamine/9009?cid=0

MSDS

https://www.chemimpex.com/product/productinfo/2-3-4-dihydro-2-i-h-i-quinolin-1-yl-ethylamine/9009?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library