Skip to Content
Merck
All Photos(1)

Key Documents

860826P

Avanti

24:1(2S-OH) Ceramide

Avanti Research - A Croda Brand (860826P)

Synonym(s):

N-(2′-(S)-hydroxynervonyl)-ᴅ-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€332.00

€332.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€332.00

About This Item

Empirical Formula (Hill Notation):
C42H81NO4
CAS Number:
Molecular Weight:
664.10
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€332.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (860826P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand (860826P)

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@@H](O)CCCCCCCCCCCC/C=C\CCCCCCCC)=O)CO

InChI

1S/C42H81NO4/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-25-27-29-31-33-35-37-41(46)42(47)43-39(38-44)40(45)36-34-32-30-28-26-24-16-14-12-10-8-6-4-2/h17-18,34,36,39-41,44-46H,3-16,19-33,35,37-38H2,1-2H3,(H,43,47)/b18-17-,36-34+/t39-,40+,41-/m0/s1

InChI key

AEJPDHWODCHVKB-NFEGVRLHSA-N

General description

Ceramide is a sphingolipid. It is made up of sphingosine linked to a fatty acid through an amide bond. Ceramide is highly hydrophobic and are present in small amounts within the cell membrane. It is an N-acyl amide derivative of sphingoid bases.[1]

Biochem/physiol Actions

Ceramide is a major secondary messenger, which regulates apoptosis and cell signaling.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860826P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide and ischemia/reperfusion injury
He X and Schuchman E H
Journal of Lipids, 2018 (2018)
The physical properties of ceramides in membranes
Alonso A and Goni F M
Annual Review of Biophysics, 47, 633-654 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service