Skip to Content
Merck
All Photos(2)

Key Documents

850699C

Avanti

08:0 PE

1,2-dioctanoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

PE(8:0/8:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H42NO8P
CAS Number:
Molecular Weight:
467.53
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850699C-10mg)
pkg of 1 × 2.5 mL (850699C-25mg)
pkg of 1 × 20 mL (850699C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850699C

concentration

10 mg/mL (850699C-10mg)
10 mg/mL (850699C-25mg)
25 mg/mL (850699C-500mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O

InChI key

UNACBKDVIYEXSL-LJQANCHMSA-N

General description

Phosphoethanolamine (PE), a lipid chaperone,[1] usually seen in the membrane leaflet is abundantly present in the mitochondria.[1]

Application

08:0 PE (1,2-dioctanoyl-sn-glycero-3-phosphoethanolamine) may be used in the preparation of liposomes for conjugating with pH sensors for live cell imaging.[2] It may also be used in the micelles for kinetic measurements studies.[3] It has been used as a phosphoethanolamine (PE) lipid species in the lipid collision cross section (CCS) calibration mixture.[4]

Biochem/physiol Actions

Phosphoethanolamine (PE) participates in phospholipid metabolism.[5] It has the ability to induce nutrient starvation tolerance. PE is capable of stimulating the development of tumor.[6] It actively participates in the production of glycosylphosphatidylinositol-anchored proteins (GPI-AP).[1]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850699C-500mg)
5 mL Clear Glass Sealed Ampule (850699C-10mg)
5 mL Clear Glass Sealed Ampule (850699C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Han et al.
The Journal of biological chemistry, 272(6), 3573-3582 (1997-02-07)
Recent genetic and structural studies have shed considerable light on the mechanism by which secretory phospholipases A2 interact with substrate aggregates. Electrostatic forces play an essential role in optimizing interfacial catalysis. Efficient and productive adsorption of the Class I bovine
Tsuyoshi Osawa et al.
Cell reports, 29(1), 89-103 (2019-10-03)
Tolerance to severe tumor microenvironments, including hypoxia and nutrient starvation, is a common feature of aggressive cancer cells and can be targeted. However, metabolic alterations that support cancer cells upon nutrient starvation are not well understood. Here, by comprehensive metabolome
Jungju Yoon et al.
Advanced materials (Deerfield Beach, Fla.), 26(26), 4559-4564 (2014-05-03)
Phase separation in films of phospholipids and conjugated polymers results in nanoassemblies because of a difference in the physicochemical properties between the hydrophobic polymers and the polar lipid heads, together with the comparable polymer side-chain lengths to lipid tail lengths
D W Ellison et al.
Brain research, 417(2), 389-392 (1987-08-11)
Measurements of both phosphoethanolamine (PEA) and ethanolamine (EA) were made in postmortem brain samples from patients with Alzheimer's disease (AD) and Huntington's disease (HD) using high-performance liquid chromatography with electrochemical detection. In AD levels of PEA were significantly reduced by
Ronja Marie Kühnel et al.
The Analyst, 144(9), 3030-3037 (2019-03-23)
The design of ion sensors has gained importance for the study of ion dynamics in cells, with fluorescent proton nanosensors attracting particular interest because of their applicability in monitoring pH gradients in biological microcompartments and reconstituted membrane systems. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service