Skip to Content
Merck
All Photos(1)

Key Documents

700115P

Avanti

5α-7,24-cholestadiene-d6

Avanti Research - A Croda Brand 700115P, powder

Synonym(s):

5α-cholesta-7,24-dien-3β-ol (d6)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€458.00

€458.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€458.00

About This Item

Empirical Formula (Hill Notation):
C27H38D6O
CAS Number:
Molecular Weight:
390.67
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

€458.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700115P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700115P

shipped in

dry ice

storage temp.

−20°C

General description

Oxygenated derivatives of cholesterol (oxysterols) present a remarkably diverse profile of biological activities, including effects on sphingolipid metabolism, platelet aggregation, apoptosis, protein prenylation, and cholesterol homeostasis.

Biochem/physiol Actions

5α-7,24-cholestadiene-d6 is a deuterated form of 5α-7,24-cholestadiene. Cholestadienes are generated from cholesterol on thermal processing. Food with cholesterol oxidation products elicit adverse effects in humans.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700115P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of Cholesterol Oxidation Products, Cholesterol Dimers and Cholestadienes After Thermal Processing of Cholesterol Standards and Butter
Derewiaka D
European Journal of Lipid Science and Technology, 121(9), 1800373-1800373 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service