Skip to Content
Merck
All Photos(1)

Key Documents

ATEH9902A8A7

AstaTech, Inc.

SOLUFLAZINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
631.4
MDL number:

Iupac Name

4-(2-(2,6-dichlorophenylamino)-2-oxoethyl)-1-(4-(4-fluorophenyl)-4-(pyridin-3-yl)butyl)piperazine-2-carboxamide dihydrochloride

Inchi Code

1S/C28H30Cl2FN5O2.2ClH/c29-23-6-1-7-24(30)27(23)34-26(37)18-35-14-15-36(25(17-35)28(32)38)13-3-5-22(20-4-2-12-33-16-20)19-8-10-21(31)11-9-19;;/h1-2,4,6-12,16,22,25H,3,5,13-15,17-18H2,(H2,32,38)(H,34,37);2*1H

InChI key

KQZLAOQGNIIITJ-UHFFFAOYSA-N

Purity

95%

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=271875

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F77968


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library