Skip to Content
Merck
All Photos(1)

Key Documents

ATEH984B2F63

AstaTech, Inc.

2,5-DIFLUOROBENZENE-1,4-DIOL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
146.09
MDL number:
Pricing and availability is not currently available.

Iupac Name

2,5-difluoro-1,4-benzenediol

Inchi Code

1S/C6H4F2O2/c7-3-1-5(9)4(8)2-6(3)10/h1-2,9-10H

InChI key

ZNHXVRDZYSYJMO-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=216719

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D93458


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library