Skip to Content
Merck
All Photos(1)

Key Documents

ATEH93EDCC0D

AstaTech, Inc.

(1R)-1-(4-CYCLOPENTYLOXYPHENYL)ETHYLAMINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
205.3
MDL number:

Iupac Name

(1R)-1-[4-(cyclopentyloxy)phenyl]ethanamine

Inchi Code

1S/C13H19NO/c1-10(14)11-6-8-13(9-7-11)15-12-4-2-3-5-12/h6-10,12H,2-5,14H2,1H3/t10-/m1/s1

InChI key

KNECCLNJJLLHAO-SNVBAGLBSA-N

Purity

95%

Physical Form

Colorless to Yellow Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=137245

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=N16450


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library