ATE316038489
AstaTech, Inc.
3,4-DIHYDRO-1H-SPIRO[NAPHTHALENE-2,4'-PIPERIDINE] HCL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3,4-dihydro-1H-spiro[naphthalene-2,4'-piperidine] hydrochloride
Inchi Code
1S/C14H19N.ClH/c1-2-4-13-11-14(6-5-12(13)3-1)7-9-15-10-8-14;/h1-4,15H,5-11H2;1H
InChI key
UHOIUTUSAFEJSO-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
97%
Shipping Temperature
Normal
Physical Form
White to Yellow Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=2216
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=50986
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.