APOH93ED5E80
Apollo Scientific Ltd
3,5-Dichloro-4-(trifluoromethyl)benzeneboronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
258.82
Iupac Name
(3,5-dichloro-4-(trifluoromethyl)phenyl)boronic acid
Inchi Code
1S/C7H4BCl2F3O2/c9-4-1-3(8(14)15)2-5(10)6(4)7(11,12)13/h1-2,14-15H
InChI key
YODWCXYMSFVDNK-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
95%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/35-dichloro-4-trifluoromethylbenzeneboronic-acid
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.