Z743791
ART® Non-Filtered Low Retention Tip
ART 200 REACH, size 200 μL, hinged rack, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
About This Item
hinged rack
Recommended Products
sterility
sterile
feature
ART 200 REACH
hinged rack
packaging
pkg of 5X768 ea
manufacturer/tradename
Molecular BioProducts 3541-05-HR
technique(s)
PCR: suitable
size
200 μL
volume
200 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Low retention non-filter tips
- Tip′s hydrophobic inner surface facilitates accurate data.
- Hinged racks allow for easy access to the tips.
- Compatible with radioactive compounds, viscous materials, PCR reagents, and more.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service