Z743009
Julabo® CORIO™ CD heating circulator with open bath
CD-B13, capacity 9 L (13 L), temperature 20-150 °C, stainless steel bath, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
Recommended Products
material
stainless steel bath
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9012413
temperature measuring range
20-150 °C
capacity
9 L (13 L)
external W × L × H
38 cm × 40 cm × 42 cm
opening W × L × H
30 cm × 18 cm × 15 cm
Looking for similar products? Visit Product Comparison Guide
General description
The Open Heating Bath Circulators of the new CORIO series feature high-quality bath tanks made of stainless steel and pump connections.
Your advantages
Cooling coil and bath cover available as optional accessories
Your advantages
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Easy pump change-over between internal and external circulation
- USB connection
- High-quality bath tanks made of stainless steel
- Integrated drain screw (except B5)
Cooling coil and bath cover available as optional accessories
Legal Information
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service