Skip to Content
Merck
All Photos(1)

Documents

Z742612

Sigma-Aldrich

Photo KitAlysis Starter Kit

Synonym(s):

photocatalysis kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.22

Quality Level

General description

Photo KitAlysis Starter Kit enables screening of 24 micro-scale simultaneous photocatalytic reactions with consistent and reproducible photon intensity. User guide is provided in the below hyperlink.
Photo KitAlysis Operating Instructions

Components:
  • Photo KitAlysis LED Controller
  • Blue LED Array (470 nm)
  • Photo KitAlysis Reaction Block
  • Torque screwdriver
  • Small screwdriver to easily remove torqued screws after reaction is complete

Features:
  • Designed and tested by synthetic chemists.
  • Controller provides repeatable milliamp selection for photon intensity
  • 0-30 mA variable LED output
  • 3 different LED options: blue (470 nm, included), green (527 nm, sold separately), and white (sold separately)
  • Non-magnetic LED base
  • Chemically resistant LED cover
  • PTFE coated cabling

Designed to be used with Photo KitAlysis High-Throughput Reaction Screening Kit (sold separately).

Best when used with KitAlysis Benchtop Inertion Box (sold separately).

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.

Protocols

Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service