Key Documents
Z741823
Extech Temperature, Humidity Datalogger
Kit with PC Interface
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
feature
CE compliant
resolution 0.1 %RH
resolution 0.1 °F/C
(WEEE Compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments 42275
parameter
-40-185 °F temperature (-40-85 °C)
size
4.9 in. × 3.8 in. × 2 in., 123.8x95.3x51mm
weight
9 oz254
General description
Compact, powerful and easy-to-use dataloggers record up to 16,000 readings. Multiple Dataloggers can be programmed and data downloaded from one single docking station. USB cable and software included to easily download collected data to your PC for further analysis.
Choose from one of the most recent versions:
Certificates of Analysis (COA)