Key Documents
Z687316
Sigma-Aldrich
Primary HEPA filter with vapor after-filter module for Aldrich® compact ductless air cleaning system
6 in. deep HEPA filter with 2 in. deep refillable absorption module with CI blend activated carbon
About This Item
UNSPSC Code:
41104919
NACRES:
NB.43
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)