Z530263
Aldrich® vacuum-transfer adapter
joint: ST/NS 19/22
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS19/22
stopcock size
2 mm
General description
PTFE stopcocks. For transfer of hazardous and sensitive reagents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC