Key Documents
Z309338
Quickfit® flat-bottom boiling flasks
capacity 100 mL, joint: ST/NS 34/35
Synonym(s):
FF100/5S
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
flat bottom flask
short
single-neck
Joint
joint: ST/NS34/35
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 5
manufacturer/tradename
Quickfit FF 100/5S
availability
not available in USA and Canada
capacity
100 mL
General description
With short neck and ground-glass socket. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)