Z281255
Laboratory brushes
Test tube, L 9 in., brush diam. × L 3/4 in. × 3 1/4 in.
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 4 ea
L
9 in.
brush diam. × L
3/4 in. × 3 1/4 in.
General description
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.