Z273295
Vionex® antimicrobial liquid soap
safe, non-toxic, and non-irritating to human skin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
47131800
NACRES:
NB.35
Recommended Products
General description
Gentle, skin-moisturizing, antimicrobial, liquid soap formulated for frequent use. Safe, nontoxic, and non-irritating to human skin. Reduces risk of the transmission of bacteria and viruses by cross contamination.
Packaging
18 oz bottle with pump sprayer.
Legal Information
Vionex is a registered trademark of Metrex Research Corporation
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH SVHC Candidate List
CAS No.
EU REACH Annex XVII (Restriction List)
CAS No.
EU REACH Annex XIV (Authorisation List)
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service