Documents
Z255912
Nalgene® 180 vacuum tubing
I.D. × O.D. 3/16 in. × 9/16 in., wall size 3/16 in.
Synonym(s):
plastic vacuum tubing
About This Item
UNSPSC Code:
41105108
packaging
pkg of 10 feet
manufacturer/tradename
Nalgene 8000-0055
parameter
35 psi max. pressure (52 °C)
70 psi max. pressure (25 °C)
I.D. × O.D.
3/16 in. × 9/16 in.
wall size
3/16 in.
General description
Transparent, nontoxic, autoclavable plastic tubing has thick walls for high vacuum.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries