Skip to Content
Merck
All Photos(2)

Documents

Z252972

Sigma-Aldrich

Enviro-Safe® thermometer

temperature -20-150 °C, immersion level, 76 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112303
NACRES:
NB.79

feature

immersion level 76 mm

manufacturer/tradename

HB Instruments B60502-0200

temperature measuring range

-20-150 °C

L

305 mm

Looking for similar products? Visit Product Comparison Guide

General description

Enviro-Safe thermometers are serialized, calibrated with NIST (NBS) traceable standards, and include a Statement of Accuracy.

White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).

Legal Information

Enviro-Safe is a registered trademark of H-B Instrument Co.

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service