Skip to Content
Merck
All Photos(1)

Documents

WHA10318489

Whatman® qualitative filter paper, Grade 470

W × L 230 mm × 305 mm, pkg of 25 ea

Synonym(s):

Whatman® qualitative filter paper, Grade 470, Whatman filter, Whatman paper, Z696048, paper filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

material

cellulose filters
sheets

feature

Grade 470

packaging

pkg of 25 ea

manufacturer/tradename

Whatman 10318489
Whatman Article No. 28414406 (US reference)

parameter

fast flow rate

W × L

230 mm × 305 mm

pore size

20-30 μm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Grade 470 has a soft surface. For gelatinous samples. Used for the absorption of culture media, as blotting paper, for electrophoresis and amino acid chromatography.

Features and Benefits

  • Soft surface.
  • For gelatinous samples.
  • The absorption of culture media, as blotting paper, for electrophoresis and amino acid chromatography.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service