Skip to Content
Merck
All Photos(1)

Key Documents

W505404

Sigma-Aldrich

p-Anisic acid

≥99%

Synonym(s):

4-Methoxybenzoic acid, p-Methoxybenzoic acid, Draconic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
508910
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
8.071

Assay

≥99%

mp

182-185 °C (lit.)

SMILES string

COc1ccc(cc1)C(O)=O

InChI

1S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ZEYHEAKUIGZSGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Faint, putrid, sweet, cadaverous undertone

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyunshun Shin et al.
Bioorganic & medicinal chemistry, 15(7), 2617-2623 (2007-02-14)
The first committed step in lipid A biosynthesis is catalyzed by uridine diphosphate-(3-O-(R-3-hydroxymyristoyl))-N-acetylglucosamine deacetylase (LpxC), a zinc-dependent deacetylase, and inhibitors of LpxC may be useful in the development of antibacterial agents targeting a broad spectrum of Gram-negative bacteria. Here, we
Jean J K Bankeu et al.
Journal of natural products, 74(6), 1370-1378 (2011-05-31)
Phytochemical investigation of the figs of Ficus mucuso led to the isolation of three new isoflavone dimer derivatives, mucusisoflavones A-C (1-3), together with 16 known compounds. Some of the isolates were tested in vitro for their inhibitory properties toward β-glucuronidase
Souichi Nakashima et al.
Bioorganic & medicinal chemistry, 18(6), 2337-2345 (2010-03-02)
The methanolic extract from the whole plants of Anastatica hierochuntica, an Egyptian herbal medicine, was found to inhibit melanogenesis in theophylline-stimulated murine B16 melanoma 4A5 cells. Among the constituents isolated, anastatin A, silybin A, isosilybins A and B, eriodictyol, luteolin
Fabrizio Carta et al.
Bioorganic & medicinal chemistry letters, 21(8), 2521-2526 (2011-03-16)
The inhibition of the β-carbonic anhydrases (CAs, EC 4.2.1.1) from the pathogenic fungi Cryptococcus neoformans (Can2) and Candida albicans (Nce103) with a series of 25 branched aliphatic and aromatic carboxylates has been investigated. Human isoforms hCA I and II were
Chao-Bin Xue et al.
Bioorganic & medicinal chemistry, 15(5), 2006-2015 (2007-01-30)
Phenoloxidase (PO), also known as tyrosinase, is a key enzyme in insect development, responsible for catalyzing the hydroxylation of tyrosine into o-diphenols and the oxidation of o-diphenols into o-quinones. Inhibition of PO may provide a basis for novel environmentally friendly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service