Skip to Content
Merck
All Photos(1)

Key Documents

W395803

Sigma-Aldrich

Phenyl acetate

≥99%

Synonym(s):

Acetic acid phenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC6H5
CAS Number:
Molecular Weight:
136.15
FEMA Number:
3958
Beilstein:
636458
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.688

Assay

≥99%

refractive index

n20/D 1.501 (lit.)

bp

196 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Oc1ccccc1

InChI

1S/C8H8O2/c1-7(9)10-8-5-3-2-4-6-8/h2-6H,1H3

InChI key

IPBVNPXQWQGGJP-UHFFFAOYSA-N

Gene Information

human ... PON1(5444)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service