Skip to Content
Merck
All Photos(1)

Documents

W365807

Sigma-Aldrich

1,4-Cineole

≥85%

Synonym(s):

1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3658
Beilstein:
104974
EC Number:
Council of Europe no.:
11225
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
3.007

biological source

synthetic

grade

Halal

reg. compliance

FDA 21 CFR 172.515

Assay

≥85%

refractive index

n20/D 1.445 (lit.)

bp

65 °C/16 mmHg (lit.)

mp

−46 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

spicy; vanilla

SMILES string

CC(C)C12CCC(C)(CC1)O2

InChI

1S/C10H18O/c1-8(2)10-6-4-9(3,11-10)5-7-10/h8H,4-7H2,1-3H3/t9-,10+

InChI key

RFFOTVCVTJUTAD-AOOOYVTPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Cineole is a volatile monoterpene cyclic ether typically found in the plant essential oils. It is phytotoxic and shows potent allelopathic property.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allelopathic effects of volatile cineoles on two weedy plant species.
Romagni JG, et al.
Journal of Chemical Ecology, 26(1), 303-313 (2000)
Patrícia Bezerra Gomes et al.
Pharmacology, biochemistry, and behavior, 96(3), 287-293 (2010-07-31)
Recent studies have shown that some monoterpenes exert anxiolytic- and depressant-like actions, however, these effects from monoterpene 1,4-cineole are still unknown. This work aimed to study the effects of 1,4-cineole in classic animal models for depression- and anxiety-like behavior, specifically
Y Asakawa et al.
Xenobiotica; the fate of foreign compounds in biological systems, 18(10), 1129-1134 (1988-10-01)
1. The metabolism of 1,4-cineole, a monoterpene ether, was studied in the rabbit. 2. Four neutral and one acidic metabolites were isolated from the urine and shown to be 9-hydroxy-1,4-cineole, 3,8-dihydroxy-1,4-cineole, 8,9-dihydroxy-1,4-cineole, 1,4-cineole-8-en-9-ol and 1,4-cineole-9-carboxylic acid.
Abdul Ahad et al.
Current drug delivery, 8(2), 213-224 (2011-01-18)
The purpose of this study was to investigate the effectiveness and mechanism(s) of percutaneous absorption of propranolol hydrochloride (PHCL) across rat and human cadaver skin using seven novel terpenes with reference to marker terpene 1,8-cineole. In-vitro skin permeation studies were
W Liu et al.
Applied and environmental microbiology, 59(11), 3889-3893 (1993-11-01)
A cytochrome P-450-dependent monooxygenase system that catalyzes the stereospecific hydroxylation of the monoterpene substrate 1,4-cineole was demonstrated in cell-free preparations of Bacillus cereus UI-1477. 1,4-Cineole hydroxylations were catalyzed by a 100,000 x g (1-h)-centrifuging soluble, hexane-inducible enzyme that activated and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service