Skip to Content
Merck
All Photos(1)

Key Documents

W341207

Sigma-Aldrich

Methyl 4-(methylthio)butyrate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2S
CAS Number:
Molecular Weight:
148.22
Flavis number:
12.06
Pricing and availability is not currently available. Contact Technical Service

Assay

≥97%

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Sulfurous, cabbage, fruity, pineapple

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service