Skip to Content
Merck
All Photos(1)

Documents

W316105

Sigma-Aldrich

Furfuryl isopropyl sulfide

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12OS
CAS Number:
Molecular Weight:
156.25
FEMA Number:
3161
Council of Europe no.:
2248
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.032

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.504 (lit.)

bp

79-80 °C/12 mmHg (lit.)

density

0.998 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CC(C)SCc1ccco1

InChI

1S/C8H12OS/c1-7(2)10-6-8-4-3-5-9-8/h3-5,7H,6H2,1-2H3

InChI key

WCHRNAKORAINOJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service