Skip to Content
Merck
All Photos(1)

Key Documents

W249408

Sigma-Aldrich

3-(2-Furyl)acrolein

≥99%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
FEMA Number:
2494
EC Number:
Council of Europe no.:
2252
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.034

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥99%

bp

100-102 °C/12 mmHg (lit.)

mp

49-55 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; spicy; nutty

storage temp.

2-8°C

SMILES string

[H]C(=O)\C([H])=C(/[H])c1ccco1

InChI

1S/C7H6O2/c8-5-1-3-7-4-2-6-9-7/h1-6H/b3-1+

InChI key

VZIRCHXYMBFNFD-HNQUOIGGSA-N

General description

3-(2-Furyl)acrolein is a heterocyclic aldehyde that can be used as a flavoring agent in the food industry.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food and Color Additives, 1, 1151-1151 (1997)
Encyclopedia of Food and Color Additives, 1, 1151-1151 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service