Skip to Content
Merck
All Photos(1)

Key Documents

W216305

Sigma-Aldrich

Isobornyl propionate

≥92%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O2
CAS Number:
Molecular Weight:
210.31
FEMA Number:
2163
EC Number:
Council of Europe no.:
412
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.131
Organoleptic:
herbaceous; floral; vanilla
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

Assay

≥92%

refractive index

n20/D 1.463 (lit.)

bp

102 °C/16 mmHg (lit.)

density

0.971 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous; floral; vanilla

SMILES string

CCC(=O)O[C@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

InChI

1S/C13H22O2/c1-5-11(14)15-10-8-9-6-7-13(10,4)12(9,2)3/h9-10H,5-8H2,1-4H3/t9-,10-,13+/m0/s1

InChI key

FAFMZORPAAGQFV-OUJBWJOFSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service