Key Documents
TEM-LC325NI
Sigma-Aldrich
Lacey Carbon Supported Nickel Grids
size 300 mesh, size 50 nm , hole size, box of 25 ×
Synonym(s):
Lacey/Carbon, 300 Mesh, Nickel, 25pk
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-LC325NI is currently not available for purchase. Contact Technical Service
packaging
box of 25 ×
size
300 mesh
50 nm , hole size
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)