Key Documents
TEM-HC200-AU-UL
Sigma-Aldrich
Ultra-Thin Holey Carbon Film Supported Gold Grids
size 200 mesh, box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-HC200-AU-UL is currently not available for purchase. Contact Technical Service
packaging
box of 25
size
200 mesh
General description
A 4-6 nm ultra-thin holey carbon film supported grids. Gold grids, 200 Mesh, 25/box
Choose from one of the most recent versions:
Certificates of Analysis (COA)