Key Documents
TEM-0250-CU
Sigma-Aldrich
Veco Speciment Grid
Copper, size 250 Square mesh , with Center Reference, vial of 100
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-0250-CU is currently not available for purchase. Contact Technical Service
packaging
vial of 100
size
250 Square mesh , with Center Reference
General description
Square Mesh with Center Reference. Copper Grids, 3.05 mm diameter, 0.8 mil thickness. Pitch 100, hole 60, bar 40 micron, 250 mesh Cu. Veco Grids are the most rigid grids available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)