Skip to Content
Merck
All Photos(2)

Documents

T54003

Sigma-Aldrich

2,4,5-Trichloroaniline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2NH2
CAS Number:
Molecular Weight:
196.46
Beilstein:
879091
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

bp

270 °C (lit.)

mp

93-95 °C (lit.)

SMILES string

Nc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C6H4Cl3N/c7-3-1-5(9)6(10)2-4(3)8/h1-2H,10H2

InChI key

GUMCAKKKNKYFEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sławomir Kasperowicz et al.
IUBMB life, 72(6), 1211-1219 (2020-03-13)
A series of chlorine-substituted benzotriazole derivatives, representing all possible substitution patterns of halogen atoms attached to the benzotriazole benzene ring, were synthetized as potential inhibitors of human protein kinase CK2. Basic ADME parameters for the free solutes (hydrophobicity, electronic properties)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service