Documents
SYNA792445
Synthware bushing adapter
female joint: ST/NS 24/40, male joint: ST/NS 45/50
Synonym(s):
Synthware A792445, glass bushing adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
female joint: ST/NS24/40
male joint: ST/NS45/50
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A792445
Related Categories
General description
Bushing adapters allow for compact assemblies of dissimilar joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.