Key Documents
SLW4542/02
Pyrex® Screwcap, bottle pourer with GL 45 screwthread
Synonym(s):
Pyrex, SciLabware 4542/02
Select a Size
1 EA
€135.00
€135.00
Estimated to ship onMay 13, 2025
Select a Size
Change View
1 EA
€135.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
€135.00
Estimated to ship onMay 13, 2025
material
blue polypropylene cap
white PTFE tube
manufacturer/tradename
4542/02
SciLabware
fitting
GL 45
General description
  • Manufactured from white PTFE and blue polypropylene
  • Aids the dispensing of liquids from bottles and flasks with a GL 45 screwthread
  • Provides greater control of flow with large capacity bottles that can be difficult to handle
  • Supplied complete with venting tube and attached PTFE cap
  • Temperature resistance to 140 deg. C
  • Autoclavable
  • Dishwasher safe
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)