Skip to Content
Merck
All Photos(2)

Documents

SLW3710/06

Pyrex® Büchner filter funnels with sintered disc

500 mL, disc diam. 95 mm, porosity grade 3

Synonym(s):

Pyrex, SciLabware 3710/06

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

manufacturer/tradename

3710/06
SciLabware

capacity

500 mL
500 mL

disc diam.

95 mm

pore size

grade 3 porosity

Looking for similar products? Visit Product Comparison Guide

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service