Skip to Content
Merck
All Photos(1)

Documents

SLW1519/04

Pyrex® Media-Lab Bottles, amber glass, with-out cap and pouring ring, with printed trace code

capacity 100 mL

Synonym(s):

Pyrex, SciLabware 1519/04

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

round amber glass bottle

Joint

Joints threaded neck (GL45)

feature

graduations
without cap
with printed trace code
without pouring ring

packaging

pack of 10 ea

manufacturer/tradename

SciLabware 1519/04

H

100 mm

body diam.

56 mm

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Manufactured from amber-coated borosilicate glass
  • UV absorbent up to 500 nm
  • Ideal for storage of light sensitive materials
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com

Features and Benefits

• Bottles only. Supplied without cap or pouring ring• Complies with ISO 4796• Manufactured from amber-coated borosilicate glass• UV absorbent up to 500 nm• Ideal for storage of light sensitive materials• Printed with Trace Code for downloadable batch certificate• The 25 ml size bottle features a glass lip and does not require a pouring ring.• Never autoclave bottles with a fully tightened screw cap. Always slacken off. Failure to follow this procedure can lead to breakage of the bottle.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service