SLW1003-5L
Pyrex® Griffin beakers, heavy duty, graduated
5000 mL
Synonym(s):
Pyrex, SciLabware 1003-5L
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
white graduation
feature
graduated
heavy duty
manufacturer/tradename
1003-5L
SciLabware
H × diam.
300 mm × 180 mm
capacity
5000 mL
General description
- Mechanically stronger than standard beakers
- Designed for longer life and increased user safety
- Reinforced rims and spout, less prone to breakage during washing etc.
- Thicker walls and heavier bases
- Manufactured from Pyrex borosilicate glass
- White graduations and marking spot
Other Notes
Browse all glass beakers
Legal Information
PYREX is a registered trademark of Corning, Inc.