SLW1000/18D
Pyrex® Griffin beakers, low form, with printed trace code
600 mL
Synonym(s):
Pyrex, SciLabware 1000/18D
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
white graduation
feature
low form
with printed trace code
manufacturer/tradename
1000/18D
SciLabware
H × diam.
125 mm × 90 mm
capacity
600 mL
General description
- Complies with ISO 3819 and DIN 12331
- Manufactured from Pyrex borosilicate glass
- Excellent thermal performance
- High resistance to chemical attack
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
- White graduations and marking spot
Other Notes
Browse all glass beakers
Legal Information
PYREX is a registered trademark of Corning, Inc.